Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-PCNA Rabbit Polyclonal Antibody
  ANTIA101680-100
New Product
undefined
Anti-PCNA Rabbit Polyclonal Antibody
  ANTIA101680-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ATLD2
  • Antigen symbol:
    PCNA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    S. cerevisiae
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P15873
  • Antigen synonyms:
    DNA polymerase delta auxiliary protein|Mutagen-sensitive 209 protein|etID36690.10|HGCN8729|MGC8367|cb16|fa28e03|fb36g03|Cyclin
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 50% Glycerol (filter sterilised, without Sodium Azide, and carrier free)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full length Saccharomyces cerevisiae PCNA protein (a.a. 1-258)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to PCNA for WB, IP and ChIP with samples derived from Saccharomyces cerevisiae.

  • Validated Applications: WB, IP, ChIP
  • Recommended Dilutions: WB: 1:1000, IP: 1:100

Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae