Anti-Fas Mouse Recombinant Antibody [clone: rB-R18]
Catalog # ANTIA314950-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:ALPS 1A
- Antigen symbol:Fas
- Clonality:Recombinant
- Clone:rB-R18
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P25445
- Antigen synonyms:APO1|APO 1 cell surface antigen|Apo1 antigen|APO 1|Apo-1 antigen|Apoptosis antigen 1|APO1 cell surface antigen|ALPS1A|Apo 1 antigen
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:38 - 50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human CD95 protein
- Tested applications:Functional studies, IHC-Fr
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rB-R18] antibody to Fas for FUNC, flow cytometry, IF and IHC-Fr with samples derived from human.
- Validated applications: FUNC, flow cytometry, IF, IHC-Fr
- Recommended dilutions: Functional Studies, flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC: Frozen: 0,5 to 1 µg/ml
Type: Primary
Antigen: Fas
Clonality: Recombinant
Clone: rB-R18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human