Anti-Integrin beta 3 Mouse Monoclonal Antibody [Clone: VIPL2]
Catalog # ANTIA85914-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:BDPLT16
- Antigen symbol:Integrin beta 3
- Clonality:Monoclonal
- Clone:VIPL2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05106
- Antigen synonyms:HPA 1|CD61 antigen|GP3A|BDPLT2|CD 61|HPA 4|GPIIIa|GT|CD61
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Unfortunately, this information is considered to be commercially sensitive
- Tested applications:IHC-Fr
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [VIPL2] antibody to Integrin beta 3 for flow cytometry, WB and IHC-Fr with samples derived from human and non-human primates.
- Validated applications: Flow cytometry, WB, IHC-Fr
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: VIPL2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates