Anti-HLA G Mouse Monoclonal Antibody [Clone: 5A6G7]
Catalog # ANTIA85877-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:B2 microglobulin
- Antigen symbol:HLA G
- Clonality:Monoclonal
- Clone:5A6G7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P17693
- Antigen synonyms:DADB-15K14.8|HLA class I molecule|HLA G3|HLA G antigen|HLA G histocompatibility antigen class I G|HLA 6.0|HLA G|HLA class I histocompatibility antigen, alpha chain G|HLA class I histocompatibility antigen alpha chain G
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:C-terminal amino acid sequence (22-mer) of soluble HLA-G5 and HLA-G6 proteins coupled to ovalbumin
- Tested applications:IHC-P, IHC-Fr, ICC
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [5A6G7] antibody to HLA G for WB, flow cytometry, IHC-P, IHC-Fr, ICC and ELISA with samples derived from human.
- Validated applications: WB, Flow cytometry, IHC-P, IHC-Fr, ICC, ELISA
- Recommended dilutions: IHC-Fr: 10 µg/ml, IHC-P: 10 µg/ml, Flow cytometry: 2 µg/ml
Type: Primary
Antigen: HLA G
Clonality: Monoclonal
Clone: 5A6G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human