Anti-HLA G Mouse Monoclonal Antibody [Clone: 87G]
ANTIA85741-100
New Product
- Antibody type:Primary
- Antigen name:B2 microglobulin
- Antigen symbol:HLA G
- Clonality:Monoclonal
- Clone:87G
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P17693
- Antigen synonyms:DADB-15K14.8|HLA class I molecule|HLA G3|HLA G antigen|HLA G histocompatibility antigen class I G|HLA 6.0|HLA G|HLA class I histocompatibility antigen, alpha chain G|HLA class I histocompatibility antigen alpha chain G
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:HLA-B27 transgenic mice were imunized with H-2 identical murine cells transfected with and expressing genes encoding HLA-G and human beta2-microglobulin
- Tested applications:IHC-Fr
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [87G] antibody to HLA G for flow cytometry, IHC-Fr and ELISA with samples derived from human.
- Validated applications: Flow cytometry, IHC-Fr, ELISA
- Recommended dilutions: Flow cytometry: 2 µg/ml, IHC-Fr: 10 µg/ml
Type: Primary
Antigen: HLA G
Clonality: Monoclonal
Clone: 87G
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human