Anti-beta Endorphin Mouse Monoclonal Antibody [Clone: B31.15]
ANTIA85629-100
New Product
- Antibody type:Primary
- Antigen name:beta Endorphin
- Clonality:Monoclonal
- Clone:B31.15
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:Proopiomelanocortin|Corticotropin lipotropin|POMC
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human beta Endorphin (full length native protein)
- Tested applications:IHC-P, IHC-Fr
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [B31.15] antibody to beta Endorphin for IHC-P and IHC-Fr with samples derived from human.
- Validated applications: IHC-P, IHC-Fr
- Recommended dilutions: IHC-P: 10 µg/ml
Type: Primary
Antigen: beta Endorphin
Clonality: Monoclonal
Clone: B31.15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human