Anti-PCNA Goat Polyclonal Antibody (Biotin)
ANTIA83562-100
New Product
- Antibody type:Primary
- Antigen name:ATLD2
- Antigen symbol:PCNA
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:DNA polymerase delta auxiliary protein|Mutagen-sensitive 209 protein|etID36690.10|HGCN8729|MGC8367|cb16|fa28e03|fb36g03|Cyclin
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-NGNIKLSQTSNVD
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human PCNA (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to PCNA (Biotin) for ELISA and WB with samples derived from human, mouse, rat and porcine.
- Validated applications: ELISA, WB
Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine