Anti-IgG Goat Polyclonal Antibody (trFluor™ Tb)
Supplier: AAT BIOQUEST
New Product
Certificates
About this item
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- Purified and quality-tested for consistent results
- Suitable for multiple applications
- Purified through affinity chromatography
This trFluor™ Tb goat anti-mouse IgG (H+L) conjugate is commonly used as a second step reagent for indirect immunofluorescent staining, when used in conjunction with primary antibodies.
Caution: Research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: trFluor™ Tb
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Specifications
- Catalog No:
- AATB16599
- AATB16792
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- trFluor™ Tb
- trFluor™ Tb
- Reactivity:
- Mouse
- Mouse
- Host:
- Goat
- Goat
- Isotype:
- IgG (H, L)
- IgG (H, L)
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- Minimal cross reaction to human, horse, rabbit and bovine
- Minimal cross reaction to human, horse, rabbit and bovine
- Storage temperature:
- Freeze (<−15 °C); Minimise light exposure
- Freeze (<−15 °C); Minimise light exposure
- Shipping temperature:
- 20
- 20