Order Entry
Slovenia
ContactUsLinkComponent
Anti-TCR C beta 1 Mouse Monoclonal Antibody [clone: JOVI.1]
Anti-TCR C beta 1 Mouse Monoclonal Antibody [clone: JOVI.1]
Catalog # ANTIA242904-100
Supplier:  Antibodies.com
New Product
undefined
Anti-TCR C beta 1 Mouse Monoclonal Antibody [clone: JOVI.1]
Catalog # ANTIA242904-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TCR C beta 1
  • Clonality:
    Monoclonal
  • Clone:
    JOVI.1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    28639
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Thymus, spleen, and mesenteric lymph nodes isolated from a mouse transgenic for human TCR V beta 3 C beta 1
  • Tested applications:
    IHC-Fr
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [JOVI.1] antibody to TCR C beta 1 for Flow Cytometry, IP and IHC-Fr with samples derived from human.

  • Validated applications: Flow cytometry, IP, IHC to Fr
  • Recommended dilutions: Flow cytometry: 2 to 6 µg/ml

Type: Primary
Antigen: TCR C beta 1
Clonality: Monoclonal
Clone: JOVI.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human