Order Entry
Slovenia
ContactUsLinkComponent
Anti-PCNA Rabbit Recombinant Antibody [clone: PCNA/8303R]
Anti-PCNA Rabbit Recombinant Antibody [clone: PCNA/8303R]
Catalog # ANTIA316871-100
Supplier:  ANTIBODIES.COM
New Product
Anti-PCNA Rabbit Recombinant Antibody [clone: PCNA/8303R]
Catalog # ANTIA316871-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATLD2
  • Antigen symbol:
    PCNA
  • Clonality:
    Recombinant
  • Clone:
    PCNA/8303R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P12004
  • Antigen synonyms:
    DNA polymerase delta auxiliary protein|Mutagen-sensitive 209 protein|etID36690.10|HGCN8729|MGC8367|cb16|fa28e03|fb36g03|Cyclin
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    36 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant full-length human PCNA protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [PCNA/8303R] antibody to PCNA for WB and IHC-P with samples derived from human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: PCNA
Clonality: Recombinant
Clone: PCNA/8303R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human