Anti-PCNA Mouse Recombinant Antibody [clone: rPCNA/8858]
ANTIA315482-100
New Product
- Antibody type:Primary
- Antigen name:ATLD2
- Antigen symbol:PCNA
- Clonality:Recombinant
- Clone:rPCNA/8858
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P12004
- Antigen synonyms:DNA polymerase delta auxiliary protein|Mutagen-sensitive 209 protein|etID36690.10|HGCN8729|MGC8367|cb16|fa28e03|fb36g03|Cyclin
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:36 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human PCNA protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant mouse monoclonal [rPCNA/8858] antibody to PCNA for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: PCNA
Clonality: Recombinant
Clone: rPCNA/8858
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human