Mouse GM-CSF ELISA Kit
Product Details & Documents
Mouse GM-CSF ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse GM-CSF in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 10 to 160 ng/L
- Sensitivity: 0,505 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse GM-CSF ELISA kit (A313733) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse GM-CSF in serum, plasma or other biological fluids. An antibody specific for GM-CSF has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the GM-CSF present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-GM-CSF antibody, which also binds the GM-CSF present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-GM-CSF antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-GM-CSF antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of GM-CSF captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of GM-CSF in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Specifications
Specifications
- Pk:96 Tests
- Description:Mouse GM-CSF ELISA kit
- Primary antibody reactivity:Mouse
- Sample type:Serum, plasma or other biological fluids
- Sample volume:40 μl
- Detection range:10 - 160 ng/L
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:1 h 30 min
- Cat. No.:ANTIA313733-96
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:0,505 ng/L
- Format:Pre-coated
- Target protein:GM-CSF
- Regulatory status:RUO



