Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Prss1/Anionic Trypsin 1
- Description:Rat Prss1/Anionic Trypsin 1 ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Rat Prss1/Anionic Trypsin 1 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Prss1/Anionic Trypsin 1. No significant cross-reactivity or interference between Prss1/Anionic Trypsin 1 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,625 - 40 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,375 ng/ml
- Regulatory status:RUO
Specifications
About this item
Rat Prss1/Anionic Trypsin 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Prss1/Anionic Trypsin 1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0,625 to 40 ng/ml
- Sensitivity: 0,375 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat Prss1 / Anionic Trypsin 1 ELISA Kit (A79664) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Prss1 / Anionic Trypsin 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Prss1 / Anionic Trypsin 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Prss1 / Anionic Trypsin 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Prss1 / Anionic Trypsin 1 Antibody, which binds the captured Prss1 / Anionic Trypsin 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Prss1 / Anionic Trypsin 1 captured in each well. The concentration of Prss1 / Anionic Trypsin 1 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.