- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Collagen III
- Description:Rat collagen III ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Rat Collagen III ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Collagen III. No significant cross-reactivity or interference between Collagen III and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:62,5 - 4000 pg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:37,5 pg/ml
- Regulatory status:RUO
Rat Collagen III ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Collagen III in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 62,5 to 4000 pg/ml
- Sensitivity: 37,5 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat Collagen III ELISA Kit (A79225) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Collagen III in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Collagen III has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Collagen III present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Collagen III Antibody, which binds the captured Collagen III present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Collagen III captured in each well. The concentration of Collagen III can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.