Order Entry
Slovenia
ContactUsLinkComponent
Rat GDF3 ELISA Kit
Rat GDF3 ELISA Kit
Catalog # ANTIA79370-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Rat GDF3 ELISA Kit
Catalog # ANTIA79370-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    GDF3
  • Description:
    Rat GDF3 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Rat GDF3 ELISA kit exhibits high specificity and excellent specificity for the detection of rat GDF3. No significant cross-reactivity or interference between GDF3 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    62,5 - 4000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    37,5 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rat GDF3 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat GDF3 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 62,5 - 4000 pg/ml
  • Sensitivity: 37,5 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat GDF3 ELISA kit (A79370) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat GDF3 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for GDF3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the GDF3 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-GDF3 Antibody, which binds the captured GDF3 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of GDF3 captured in each well. The concentration of GDF3 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.