Human TRP2/DCT ELISA Kit
Human TRP2/DCT ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TRP2/DCT in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0,313 to 20 ng/ml
- Sensitivity: 0,188 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human TRP2 / DCT ELISA Kit (A79262) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human TRP2 / DCT in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for TRP2 / DCT has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TRP2 / DCT present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-TRP2 / DCT Antibody, which binds the captured TRP2 / DCT present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of TRP2 / DCT captured in each well. The concentration of TRP2 / DCT can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.
: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
- Pk:96 Tests
- Description:Human TRP2/DCT ELISA kit
- Primary antibody reactivity:Human
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Sample volume:100 μl
- Detection range:0,313 - 20 ng/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:4 h 30 min
- Cross reactivity:Human TRP2/DCT ELISA Kit exhibits high specificity and excellent specificity for the detection of human TRP2/DCT. No significant cross-reactivity or interference between TRP2/DCT and analogues was observed.
- Assay Principle:Quantitative
- Cat. No.:ANTIA79262-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:0,188 ng/ml
- Format:Pre-coated
- Target protein:TRP2/DCT
- Regulatory status:RUO
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



