Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Human Stanniocalcin 2/STC-2 ELISA Kit

Human Stanniocalcin 2/STC-2 ELISA Kit

Human stanniocalcin 2/STC-2 ELISA kit, 96 Tests
Catalog # ANTIA78829-96
Supplier:   Antibodies.com
Human Stanniocalcin 2/STC-2 ELISA Kit
Catalog # ANTIA78829-96
Supplier:  Antibodies.com
CAS Number:  
Human Stanniocalcin 2/STC-2 ELISA Kit
Human Stanniocalcin 2/STC-2 ELISA Kit

Human stanniocalcin 2/STC-2 ELISA kit, 96 Tests

CAS Number:  
Price
625,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human Stanniocalcin 2/STC-2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Stanniocalcin 2/STC-2 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 31,25 - 2000 pg/ml
  • Sensitivity: 18,75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human Stanniocalcin 2/STC-2 ELISA kit (A78829) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Stanniocalcin 2/STC-2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Stanniocalcin 2/STC-2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Stanniocalcin 2/STC-2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Stanniocalcin 2/STC-2 Antibody, which binds the captured Stanniocalcin 2/STC-2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Stanniocalcin 2/STC-2 captured in each well. The concentration of Stanniocalcin 2/STC-2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Human stanniocalcin 2/STC-2 ELISA kit
  • Primary antibody reactivity:
    Human
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    31,25 - 2000 pg/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Human Stanniocalcin 2/STC-2 ELISA kit exhibits high specificity and excellent specificity for the detection of human Stanniocalcin 2/STC-2. No significant cross-reactivity or interference between Stanniocalcin 2/STC-2 and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    ANTIA78829-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    18,75 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Stanniocalcin 2/STC-2
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.