Order Entry
Slovenia
ContactUsLinkComponent
Human Complement C8a ELISA Kit
Human Complement C8a ELISA Kit
Catalog # ANTIA79158-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human Complement C8a ELISA Kit
Catalog # ANTIA79158-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Complement C8a
  • Description:
    Human complement C8a ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Human Complement C8a ELISA Kit exhibits high specificity and excellent specificity for the detection of human Complement C8a. No significant cross-reactivity or interference between Complement C8a and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    12,5 - 800 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    7,5 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human Complement C8a ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Complement C8a in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 12,5 to 800 ng/ml
  • Sensitivity: 7,5 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Complement C8a ELISA Kit (A79158) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Complement C8a in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement C8a has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement C8a present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Complement C8a Antibody, which binds the captured Complement C8a present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement C8a captured in each well. The concentration of Complement C8a can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.