Order Entry
Slovenia
ContactUsLinkComponent
Human Neurotensin ELISA Kit
Human Neurotensin ELISA Kit
Catalog # ANTIA79576-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human Neurotensin ELISA Kit
Catalog # ANTIA79576-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Neurotensin
  • Description:
    Human neurotensin ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Human Neurotensin ELISA kit exhibits high specificity and excellent specificity for the detection of human Neurotensin. No significant cross-reactivity or interference between Neurotensin and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1,563 - 100 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    0,938 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human Neurotensin ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human Neurotensin in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 1,563 - 100 pg/ml
  • Sensitivity: 0,938 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human Neurotensin ELISA kit (A79576) employs the competitive enzyme immunoassay technique for the quantitative measurement of human Neurotensin in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Neurotensin antigen. During the incubation, Neurotensin present in the samples or standards competes with the fixed amount of immobilized Neurotensin for binding sites on the Biotinylated Anti-Neurotensin Antibody. The more Neurotensin present in a sample or standard, the less Biotinylated Anti-Neurotensin Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Neurotensin Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Neurotensin present in each sample or standard. The concentration of Neurotensin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.