- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Mu Opioid receptor
- Description:Rat Mu Opioid receptor ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Rat Mu Opioid receptor ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Mu Opioid receptor. No significant cross-reactivity or interference between Mu Opioid receptor and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:31,25 - 2000 pg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:18,75 pg/ml
- Regulatory status:RUO
Rat Mu Opioid receptor ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Mu Opioid receptor in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 31,25 to 2000 pg/ml
- Sensitivity: 18,75 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat Mu Opioid Receptor ELISA Kit (A79593) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Mu Opioid Receptor in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Mu Opioid Receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Mu Opioid Receptor present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Mu Opioid Receptor Antibody, which binds the captured Mu Opioid Receptor present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Mu Opioid Receptor captured in each well. The concentration of Mu Opioid Receptor can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.