VIC Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Catalog # AATB67002
Supplier: AAT Bioquest
New Product
Specifications
- Frame colour:White
- No. of wells:96
- Profile:Low
- Skirt:Semi-skirted
- Well colour:White
- Description:VIC Dye qPCR calibration plate
- Barcoded:Yes
- Pk:1
Specifications
About this item
The VIC dye qPCR calibration plate can be used to maintain the 7500 real-time PCR system with fast 96-well block.
- The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
- This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Caution: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3 times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimise light exposure.