ContactUsLinkComponent
DrägerSensor® Electrochemical Sensors
DrägerSensor® Electrochemical Sensors
Supplier:  Dräger
New Product
Certificates
DrägerSensor® Electrochemical Sensors
Supplier:  Dräger
New Product
Specifications
  • Catalog No:
  • DRAE6809605
  • DRAE6809685
  • Accuracy:
  • ≤1%
  • ≤3%
  • Temperature range:
  • −40...+65 °C
  • −40...+65 °C
  • Measurement range:
  • 50 - 1000 ppm
  • 500 - 3000 ppm