About this item
Mouse monoclonal IgG1 antibody against human MUC1 (CD227), PE-conjugated.
- Airway cells
The 16A antibody reacts with human MUC1, a large (>250 kDa) heavily glycosylated type 1 transmembrane protein expressed on the surface of most glandular and ductal epithelial cells and a variety of hematopoietic cells. A characteristic feature of the MUC1 glycoprotein is a core domain composed of a variable number of tandem repeats and multiple oligosaccharide side chains. Because the extracellular portion of MUC1 can extend beyond most cell surface proteins, it is thought to play a role in cell-cell and cell-substrate adhesion. The protein is highly expressed by a majority of human adenocarcinomas and is associated with a poor prognosis. In the mammary gland, MUC1 is localized on the apical plasma membrane of luminal epithelial cells. The clone 16A antibody has a higher affinity for the glycosylated form of MUC1.
Type:
Antigen: MUC1 (CD227)
Clonality: Monoclonal
Clone: 16A
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1 lambda
Reactivity: Human
Specifications
- Catalog No:
- STMC60155PE.1
- STMC60155PE
- Antigen name:
- MUC1 (CD227)
- MUC1 (CD227)
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 16A
- 16A
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 4582
- 4582
- Isotype:
- IgG1 lambda
- IgG1 lambda