Order Entry
Slovenia
ContactUsLinkComponent
Anti-CD33 Mouse Monoclonal Antibody [clone: HIM3-4] (PE)
Anti-CD33 Mouse Monoclonal Antibody [clone: HIM3-4] (PE)
Certificates
undefined
Anti-CD33 Mouse Monoclonal Antibody [clone: HIM3-4] (PE)

Specifications

  • Catalog No:
  • STMC60096PE
  • STMC60096PE.1
  • Antigen name:
  • CD33
  • CD33
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • HIM3-4
  • HIM3-4
  • Reactivity:
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 945
  • 945
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together