Order Entry
Slovenia
ContactUsLinkComponent
Anti-SSEA-5 Mouse Monoclonal Antibody [clone: 8E11]
Anti-SSEA-5 Mouse Monoclonal Antibody [clone: 8E11]
Certificates
undefined
Anti-SSEA-5 Mouse Monoclonal Antibody [clone: 8E11]

Specifications

  • Catalog No:
  • STMC60063.1
  • STMC60063
  • Antigen name:
  • SSEA-5
  • SSEA-5
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 8E11
  • 8E11
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together