TISSUE-TROL™ Control Slides Helicobacter
Catalog # TTR006-25EA
Supplier: SIGMA ALDRICH MICROSCOPY
Specifications
- Species:
- Description:Helicobacter TISSUE-TROL™ control slides from mouse intestine (tissue containing helicobacter pylori)
- Environmentally Preferable:
- Pk:25
Specifications
About this item
Histology slides with paraffin embedded section of mouse intestine tissue containing Helicobacter pylori. Suitable for use with modified steiner-steiner silver stain procedure No. HT101.
Delivery: Box contains 24 unstained slides plus one reference slide.