Quick Clip
Quick clip, Polyamide, 220 mm, White
Catalog # SAMP8219A-220
Supplier: Sampling Systems
Jump to:
Product Details & Documents
Quick clip provides an instant seal for bags.
- Provides an instant powder- and water-tight seal for bags
Quick clips can be used over and over again, providing a convenient and cost-eefective solution to sealing different types of bags.
Specifications
- Colour:White
- Description:Quick clip
- Length:220 mm
- Material:Polyamide