Specifications
- Pk:50 mL
- Description:IntestiCult™ OGM Human basal medium
Specifications
About this item
Cell culture medium for establishment and maintenance of human colorectal cancer organoids.
Efficiently grow Wnt-independent colorectal tumor organoids using IntestiCult™ OGM Human Basal Medium. Tumoroids have high predictive value as an in vitro system as they retain the characteristics of the tumor of origin. Use IntestiCult™ OGM Human Basal Medium to provide optimal culture conditions for tumor organoid growth and expansion while maintaining tumor heterogeneity. The Wnt-free formulation ensures that Wnt-dependent, normal, healthy intestinal cells do not contaminate the tumoroid culture.
Tumor organoids generated using IntestiCult™ OGM Human Basal Medium are suitable for a variety of applications, including modeling tumor development and screening drugs for efficacy and toxicity. For growing healthy intestinal organoids, please use IntestiCult™ Organoid Growth Medium (Human) or IntestiCult™-SF Organoid Growth Medium (Human). For differentiating human intestinal organoids, refer to IntestiCult™ Organoid Differentiation Medium (Human).