ContactUsLinkComponent
MesenCult™ Proliferation Kit (Human)
MesenCult™ Proliferation Kit (Human)
Catalog # STMC05411
CAS Number:  
undefined
MesenCult™ Proliferation Kit (Human)
Catalog # STMC05411
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    MesenCult™ Proliferation kit (Human)
Specifications
About this item
Medium for detection of CFU-F and expansion of human mesenchymal stem cells.
MesenCult™ Proliferation Kit (Human) is a standardized, serum-containing medium for the culture of human mesenchymal stem cells (MSCs). MesenCult™ Proliferation Kit (Human) is optimised for the expansion of human MSCs in vitro as well as for the detection and enumeration of colony-forming unit - fibroblasts (CFU-F). MesenCult™ Proliferation Kit (Human) includes MesenCult™ MSC Basal Medium (Human; 450 ml) and MesenCult™ MSC Stimulatory Supplement (Human; 50 ml).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').