ContactUsLinkComponent
IntestiCult™-SF Organoid Growth Medium (Human)
IntestiCult™-SF Organoid Growth Medium (Human)
Catalog # STMC100-0340
CAS Number:  
undefined
IntestiCult™-SF Organoid Growth Medium (Human)
Catalog # STMC100-0340
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    IntestiCult™ SF organoid growth medium (human)
Specifications
About this item
Serum-free and conditioned medium-free cell culture medium for establishment and maintenance of human intestinal organoids.
IntestiCult™-SF Organoid Growth Medium (Human) is a complete cell culture medium for efficient establishment and long-term maintenance of organoids derived from human intestinal crypts. IntestiCult™-SF is a serum-free and conditioned medium-free formulation. Intestinal organoids provide a convenient in vitro organotypic culture system for studying the intestinal epithelium. Isolated intestinal crypts rapidly form complex organoids when cultured in IntestiCult™-SF. The organoids incorporate a functional lumen enclosed by a polarised epithelial cell layer containing known cell types of the adult intestinal epithelium. Maturing intestinal organoids establish protruding buds, resembling intestinal crypts of the in vivo tissues.
IntestiCult™-SF supports the proliferation of intestinal epithelial stem cells from patient’s biopsies and expansion of organoid cultures derived from fresh tissue or from already established frozen stocks. Applications of intestinal organoid cultures include studying the development and function of intestinal epithelium, modeling intestinal diseases, and performing targeted molecule screens. Intestinal organoid cultures can also be used for investigation of adult stem cell properties and for regenerative therapy approaches, and can be further differentiated using IntestiCult™ Organoid Differentiation Medium.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').