Order Entry
Slovenia
ContactUsLinkComponent
Anti-MAGEA1 Mouse Monoclonal Antibody [clone: SPM282]
Anti-MAGEA1 Mouse Monoclonal Antibody [clone: SPM282]
  PRSI34-131
Anti-MAGEA1 Mouse Monoclonal Antibody [clone: SPM282]
  PRSI34-131

 

  • Antibody type:
    Primary
  • Antigen name:
    melanoma antigen family A, 1 (directs expression of antigen MZ2-E)
  • Antigen symbol:
    MAGEA1
  • Clonality:
    Monoclonal
  • Clone:
    SPM282
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Dog,
    Rat
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    P43355
  • Antigen synonyms:
    MAGEA1
  • Storage buffer:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Storage temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    Full length human protein was used as the immunogen for this MAGE-1 antibody.
  • Purification:
    Protein G affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recognises a protein of 42-46 kDa, identified as MAGE-1. This mAb does not cross-react with MAGE-2, -3, -4, -6 -9, -10, -or -12 protein. Human malignant neoplasms carry rejection antigens that are recognized by the patients' autologous, tumor directed and specific, cytolytic, CD8+ T lymphocyte clones (CTL). The MAGE family of genes codes an important group of antigens. It was identified that melanomas and primary glial brain tumors express common melanoma associated antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various histological types (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy.

Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the MAGE-1 antibody for each application should be determined by the researcher.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: MAGEA1
Clonality: Monoclonal
Clone: SPM282
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Rat, Dog