Specifications
- Antibody type:Primary
- Antigen name:keratin 6A, keratin 6B
- Antigen symbol:KRT6A, KRT6B
- Clonality:Monoclonal
- Clone:SPM269
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P02538, P04259, P48668
- Antigen synonyms:KRT6A, KRT6B
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Amino acids GSSTIKYTTTS from the C-terminus of human keratin 6 was used as the immunogen for this Cytokeratin 6 antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
In humans, multiple isoforms of Cytokeratin 6 (6A-6F), encoded by several highly homologous genes, have distinct tissue expression patterns, and Cytokeratin 6A is the dominant form in epithelial tissue. The gene encoding human Cytokeratin 6A maps to chromosome 12q13, and mutations in this gene are linked to several inheritable hair and skin pathologies. Keratins 6 and 16 are expressed in keratinocytes, which are undergoing rapid turnover in the suprabasal region (also known as hyper-proliferation-related keratins). Keratin 6 is found in hair follicles, suprabasal cells of a variety of internal stratified epithelia, in epidermis, in both normal and hyper-proliferative situations. Epidermal injury results in activation of keratinocytes, which express CK6 and CK16. CK6 is strongly expressed in about 75% of head and neck squamous cell carcinomas. Expression of CK6 is particularly associated with differentiation.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,1-0,2 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the Cytokeratin 6 antibody for each application should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: KRT6A, KRT6B
Clonality: Monoclonal
Clone: SPM269
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Mouse