Order Entry
Slovenia
ContactUsLinkComponent
Anti-NKX2-1 Mouse Monoclonal Antibody [clone: NX2.1/690]
Anti-NKX2-1 Mouse Monoclonal Antibody [clone: NX2.1/690]
Catalog # PRSI33-470
Supplier:  ProSci Inc.
Anti-NKX2-1 Mouse Monoclonal Antibody [clone: NX2.1/690]
Catalog # PRSI33-470
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NK2 homeobox 1
  • Antigen symbol:
    NKX2-1
  • Clonality:
    Monoclonal
  • Clone:
    NX2.1/690
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    P43699
  • Antigen synonyms:
    NKX2-1
  • Storage buffer:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Storage temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    Recombinant protein was used as the immunogen for the TTF1 antibody.
  • Purification:
    Protein G affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recognises a protein of 40 kDa, identified as Thyroid transcription factor-1. TTF-1 is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. Anti-TTF-1 is useful in differentiating primary adenocarcinoma of the lung from metastatic carcinomas originating in the breast, mediastinal germ cell tumors, and malignant mesothelioma. It can also be used to differentiate small cell lung carcinoma from lymphoid infiltrates. Loss of TTF-1 expression in non-small cell lung carcinoma has been associated with aggressive behavior of such neoplasms. TTF-1 reactivity is also seen in thyroid malignancies.

Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Optimal dilution of the TTF1 antibody should be determined by the researcher.

1. Staining of formalin/paraffin tissues requires boiling tissue sections in 10 mM Citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 min.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: NKX2-1
Clonality: Monoclonal
Clone: NX2.1/690
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse, Rat