Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) receptor 5
- Antigen symbol:CCR5
- Clonality:Monoclonal
- Clone:12D1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Neutralisation:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P51681
- Antigen synonyms:CCR5
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Human native protein was used as the immunogen for the CCR5 antibody.
- Tested applications:Functional
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Reacts with the N-terminal extracellular domain of CD195. The CC chemokine receptor 5 (CCR5) is a member of the CC-chemokine receptor family, and has the characteristic structure of a 7 transmembrane G protein-coupled receptor (GPCR). CCR5 regulates trafficking and effector functions of memory/effector Th1 cells, macrophages, NK cells, and immature dendritic cells. CCR5 and its ligands play an important role in viral pathogenesis. CCR5 represents the co-receptor for macrophage (M) and dual (T cell and M)-tropic immunodeficiency viruses. Together with the CD4 binding receptor, CCR5 plays a critical role in HIV entry into the target cells. Moreover, the CCR5 ligands macrophage inflammatory protein (MIP)-1 alpha, MIP-1 beta and RANTES act as endogenous inhibitors of HIV infection, making both CCR5 and its chemokine ligands attractive therapeutic targets for HIV infection. Recent studies have also highlighted the role of CCR5 in a variety of other human diseases, ranging from infectious and inflammatory diseases to cancer.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Functional Activity (Order SAF formulation) Immunohistochemistry (Frozen): 0,5 - 1 µg/ml for 30 min at RT Prediluted format: incubate for 30 min at RT (2) Optimal dilution of the CCR5 antibody should be determined by the researcher.
1. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: 12D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human