Specifications
- Antibody type:Primary
- Antigen name:keratin 1
- Antigen symbol:KRT1
- Clonality:Monoclonal
- Clone:34BE12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P04264
- Antigen synonyms:KRT1
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Solubilized keratin extract from human stratum corneum was used as the immunogen for this HMW Cytokeratin antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This antibody recognises HMW Keratin 1, 5, 10 and 14. In normal epithelia, it stains stratified epithelia, myoepithelial cells and basal cells in the prostate gland and bronchi. The HMW Cytokeratin antibody shows no reactivity with hepatocytes, pancreatic acinar cells, proximal renal tubules, or endometrial glands; there is no reactivity with cells derived from simple epithelia. Mesenchymal tumors, lymphomas, melanomas, neural tumors, and neuroendocrine tumors are negative with this mAb. It stains myoepithelial cells and has been shown to be useful in distinguishing prostate adenocarcinoma from benign prostate. It has also been useful in separating benign from malignant intraductal breast proliferations.
Flow Cytometry: 0,5 - 1 µg/10e6 cells IF: 0,5-1,0 µg/ml WB: 0,5-1,0 µg/ml IHC (FFPE): 0,5-1,0 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the HMW Cytokeratin antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: KRT1
Clonality: Monoclonal
Clone: 34BE12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse, Rat