Anti-CYP1B1 Rabbit Polyclonal Antibody
Catalog # PRSI15-298
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450, family 1, subfamily B, polypeptide 1
- Antigen symbol:CYP1B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1545
- Antigen synonyms:CYPIB1|CYP1B1|CP1B|P4501B1|GLC3A
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 244-543 of human CYP1B1 (NP_000095.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CYP1B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse