Anti-ATP1A2 Rabbit Polyclonal Antibody
PRSI15-211
New Product
- Antibody type:Primary
- Antigen name:ATPase Na+/K+ transporting subunit alpha 2
- Antigen symbol:ATP1A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:477
- Antigen synonyms:sodium pump subunit alpha-2|sodium/potassium-transporting ATPase subunit alpha-2|FHM2|Na+/K+ ATPase|MHP2|sodium/potassium-transporting ATPase alpha-2 chain|alpha-A(+) catalytic polypeptide|Na(+)/K(+) ATPase alpha-2 subunit|alpha-B polypeptide|sodium-potassium ATPase catalytic subunit alpha-2|ATPase Na+/K+ transporting alpha 2 polypeptide
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human ATP1A2 (NP_000693.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: ATP1A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat