Anti-EDC3 Rabbit Polyclonal Antibody
PRSI15-190
- Antibody type:Primary
- Antigen name:Enhancer of mRNA-decapping protein 3
- Antigen symbol:EDC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:80153
- Antigen synonyms:hYjeF_N2|Enhancer of mRNA-decapping protein 3|YJDC|YjeF N-terminal domain-containing protein 2|LSM16|LSM16 homolog|EDC3|YJEFN2|YjeF_N2|YjeF domain-containing protein 1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 159-508 of human EDC3 (NP_079359.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: EDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat