Anti-KLHDC3 Rabbit Polyclonal Antibody
PRSI15-179
New Product
- Antibody type:Primary
- Antigen name:Kelch domain containing 3
- Antigen symbol:KLHDC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:116138
- Antigen synonyms:dJ20C7.3|testis intracellular mediator protein|PEAS|kelch domain-containing protein 3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-382 of human KLHDC3 (NP_476502.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: KLHDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat