Anti-CD1D Rabbit Polyclonal Antibody
PRSI15-010
- Antibody type:Primary
- Antigen name:CD1d molecule
- Antigen symbol:CD1D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:912
- Antigen synonyms:CD1D|Thymocyte antigen CD1D|R3|CD1D antigen|d polypeptide|CD1d molecule|R3G1|CD1d antigen
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 20-190 of human CD1D (NP_001757.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: CD1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse