Anti-Snap23 Rabbit Monoclonal Antibody [clone: A6]
Catalog # PRSI54-339
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:synaptosomal-associated protein 23
- Antigen symbol:Snap23
- Clonality:Monoclonal
- Clone:A6
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:20619
- Antigen synonyms:Sndt|23kDa|AA408749|Syndet|SNAP-23
- Modification:None
- Storage buffer:PBS, pH 7,2, 0,02% NaN₃
- Storage temperature:Store at 4 °C for up to 6 months. Store at −20 °C long term
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Synthetic peptide derived from mouse Snap23 (CRTKNFESGKNYKA)
- Purification:Protein A purified
- Pack type:Vial
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Snap23
Clonality: Monoclonal
Clone: A6
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse