Anti-UBAP2 Rabbit Polyclonal Antibody
Catalog # PRSI56-637
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin-associated protein 2
- Antigen symbol:UBAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Amino acid range = 144-173
- Form:Liquid
- Gene ID:Q5T6F2
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:117 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This UBAP2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 144-173 amino acids from the N-terminal region of human UBAP2
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
This gene is a novel gene isolated based on its expression in the human adrenal gland. The full-length protein encoded by this gene contains a UBA-domain (ubiquitin associated domain), which is a motif found in several proteins having connections to ubiquitin and the ubiquitination pathway. In addition, the protein contains a region similar to a domain found in members of the atrophin-1 family. The function of this protein has not been determined. Additional alternate splice variants may exist, but their full length nature has not been determined.
- For WB starting dilution is: 1:1000
- For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: UBAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Mouse