Orders LinkContactUsLinkComponent
UST bench brush, soft, Vikan®
UST bench brush, soft, Vikan®
Supplier:   Basan
UST bench brush, soft, Vikan®
Supplier:  Basan
Certificates
UST bench brush, soft, Vikan®
UST bench brush, soft, Vikan®
8 Options Available Below
Product Details & Documents
This ULTRA SAFE TECHNOLOGY (UST) Bench Brush with Ultra Filament Security ensures effective sweeping of fine dry particles from conveyor belts, food preparation surfaces, tables and equipment in high-risk areas.
  • Soft bristles made from Polybutylene terephthalate (PBT)
  • Handle made from glass reinforced PP
  • Large choice of colours
  • Autoclavable at 121 °C
All UST brushes feature a unique bristle system that minimises the risk of contamination and bristle loss.
Certifications: Food contact approved. FDA compliant.
Product Family Options
Product Information
  • Bristle material
    Bristle colour
    Total length
    Handle colour
    Trim length
    Availability
    Price
  • PBT
    Pink
    330 mm
    Pink
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,10 €
    27,10 €
    1 * 1 items
       27,10 €
  • PBT
    Green
    330 mm
    Green
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,10 €
    27,10 €
    1 * 1 items
       27,10 €
  • PBT
    Blue
    330 mm
    Blue
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,10 €
    27,10 €
    1 * 1 items
       27,10 €
  • PBT
    Red
    330 mm
    Red
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,10 €
    27,10 €
    1 * 1 items
       27,10 €
  • PBT
    White
    330 mm
    White
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,10 €
    27,10 €
    1 * 1 items
       27,10 €
  • PBT
    Yellow
    330 mm
    Yellow
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,10 €
    27,10 €
    1 * 1 items
       27,10 €
  • PBT
    Orange
    330 mm
    Orange
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,10 €
    27,10 €
    1 * 1 items
       27,10 €
  • PBT
    Purple
    330 mm
    Purple
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 27,10 €
    27,10 €
    1 * 1 items
       27,10 €