Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-ω-Propyl-L-arginine
N-ω-Propyl-L-arginine ≥98%
N-ω-Propyl-L-arginine ≥98%
N-ω-Propyl-L-arginine ≥98%
Catalog # ENZOALX270203M025
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
N-ω-Propyl-L-arginine ≥98%
Catalog # ENZOALX270203M025
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
CAS Number:
137361-05-8
Product Details