ContactUsLinkComponent
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥95% (by HPLC), Ultrapure dye
Catalog # ENZOENZ52401
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
undefined
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥95% (by HPLC), Ultrapure dye
Catalog # ENZOENZ52401
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Specifications
  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:
Specifications
About this item
Synonyms: Hoechst 33342, 2′-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride, HOE 33342, 2'-(4-Ethoxyphenyl)-5-(4-methylpiperazin-1-yl)-1H,1'H-2,5'-bibenzo[d]imidazole, bisBenzimide
 :
 :561.93 g/mol
Storage Temperature:Freezer
CAS Number:23491-52-3
 :MFCD00012679
Formula:C₂₇H₂₈N₆O·3HCl