Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
STA-21
STA-21 ≥98% (by TLC)
STA-21 ≥98% (by TLC)
STA-21 ≥98% (by TLC)
Catalog # ENZOBMLEI3510001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
STA-21 ≥98% (by TLC)
Catalog # ENZOBMLEI3510001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
CAS Number:
28882-53-3
Product Details