0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
U-75302
U-75302 ≥98% (by TLC)
U-75302 ≥98% (by TLC)
ENZOBMLRA1020100
:
ENZO LIFE SCIENCES
:
U-75302 ≥98% (by TLC)
ENZOBMLRA1020100
:
ENZO LIFE SCIENCES
:
Pk:
100 µG
Seq:
1286
Purity:
≥98% (TLC)
:
:
361.5 g/mol
:
GHS07
:
Ultra-Low Temperature Freezer (–80°C)
:
119477-85-9
:
C₂₂H₃₅NO₃