Order Entry
Slovenia
ContactUsLinkComponent
 
1-O-Hexadecyl-2-O-acetyl-sn-glycero-3-phosphocholine ≥98% (by TLC)
  ENZOBMLL1000005
 :  
undefined
1-O-Hexadecyl-2-O-acetyl-sn-glycero-3-phosphocholine ≥98% (by TLC)
  ENZOBMLL1000005
 :  

 

  • Pk:
    5 mg
  • Seq:
    2210

 

 

  • Purity:
    ≥98% (TLC)
  • Identity:
    Determined by IR

 

 

 :
 :523.69 g/mol
 :14,07524
 :GHS07
 :GHS08
 :Freezer
 :74389-68-7
 :C₂₆H₅₄NO₇P
 :MFCD00043214