C2-Dihydroceramide ≥98% (by TLC)
Catalog # ENZOBMLSL1010005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Purity:≥98% (TLC)
Specification Test Results
About this item
:
:343.552 g/mol
Storage Temperature:Freezer
CAS Number:13031-64-6
Formula:C₂₀H₄₁NO₃
MDL Number:MFCD00236495