Order Entry
Slovenia
ContactUsLinkComponent
2,3-Bis(4-hydroxyphenyl)propionitrile ≥98% (by TLC), white solid
2,3-Bis(4-hydroxyphenyl)propionitrile ≥98% (by TLC), white solid
Catalog # ENZOBMLGR2480050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
2,3-Bis(4-hydroxyphenyl)propionitrile ≥98% (by TLC), white solid
Catalog # ENZOBMLGR2480050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

 :
 :239.27 g/mol
 :GHS07
 :GHS09
Storage Temperature:Freezer
CAS Number:1428-67-7
Formula:C₁₅H₁₃NO₂
MDL Number:MFCD01695412